missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanecarboxylic acid chloride, 97%

Product Code. 10081840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081840 25 g Glass bottle 25g
10754081 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081840 Supplier Thermo Scientific Acros Supplier No. 152480250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2094-72-6 | C11H15ClO | 198.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2094-72-6
Molecular Formula C11H15ClO
Molecular Weight (g/mol) 198.69
MDL Number MFCD00074724
InChI Key MIBQYWIOHFTKHD-UHFFFAOYSA-N
Synonym 1-adamantanecarbonyl chloride, adamantanecarbonyl chloride, 1-adamantanecarbonylchloride, 1-adamantoyl chloride, adamantoyl chloride, 1-adamantanoic acid chloride, 1-adamantanecarboxylic acid chloride, adamantane-1-carboxylic acid chloride, 1-adamantylcarbonyl chloride, 1-adamantyl carbonyl chloride
PubChem CID 98915
IUPAC Name adamantane-1-carbonyl chloride
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color White
Boiling Point 135.0°C to 136.0°C (10.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Fieser 02,15
Solubility Information Solubility in water: may decompose
Formula Weight 198.69
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 1-Adamantanecarboxylic acid chloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.