missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetonitrile, 97%

Product Code. 11476496
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476496 5 g 5g
11466496 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476496 Supplier Thermo Scientific Alfa Aesar Supplier No. B24770.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16269-13-9 | C12H17N | 175.28 g/mol

1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 16269-13-9
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00194034
InChI Key DXQVPXCZIRQITG-UHFFFAOYSA-N
Synonym 2-adamantan-1-yl acetonitrile, 1-adamantaneacetonitrile, 1-adamantylacetonitrile, 2-1-adamantyl acetonitrile, tricyclo 3.3.1.13,7 decane-1-acetonitrile, 2-adamantanylethanenitrile, pubchem21521, 1-adamantylmethyl cyanide, adamantane-1-acetonitrile
PubChem CID 140052
SMILES N#CCC12CC3CC(CC(C3)C1)C2

Spezifikation

Quantity 5 g
UN Number UN3439
Solubility Information Sparingly soluble in water.
IUPAC Name 2-(adamantan-1-yl)acetonitrile
Formula Weight 175.28
Percent Purity 97%
Chemical Name or Material 1-Adamantaneacetonitrile

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.