missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetic acid, 98+%

Product Code. 11388565
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388565 1 g 1g
11398565 5 g 5g
11308575 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388565 Supplier Thermo Scientific Alfa Aesar Supplier No. L12029.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4942-47-6 | C12H17O2 | 193.27 g/mol

1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Solubility
Solubility in Methanol - almost transparency

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4942-47-6
Molecular Formula C12H17O2
Molecular Weight (g/mol) 193.27
MDL Number MFCD00074728
InChI Key AOTQGWFNFTVXNQ-UHFFFAOYSA-M
Synonym 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
PubChem CID 123221
IUPAC Name 2-(1-adamantyl)acetic acid
SMILES [O-]C(=O)CC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 136°C to 138°C
Quantity 1 g
Beilstein 641412
Formula Weight 194.28
Percent Purity ≥98%
Chemical Name or Material 1-Adamantaneacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.