missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine, 98%

Product Code. 15423867
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423867 100 g 100g
15403867 5 g 5g
15413867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423867 Supplier Thermo Scientific Alfa Aesar Supplier No. H30076.22
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 768-94-5 | C10H17N | 151.253 g/mol

1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N

Specifications

Melting Point ∼200°C (sublimation)
Quantity 100 g
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 151.25
Percent Purity 98%
Chemical Name or Material 1-Adamantanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.