missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine, 96%

Product Code. 10142764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10142764 5 g Glass bottle 5g
10051694 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142764 Supplier Thermo Scientific Acros Supplier No. 160470050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 768-94-5 | C10H17N | 151.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.25
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N

Specifications

CAS Min % 95
CAS Max % 100
Melting Point 208.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 0.96
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 368
Solubility Information Solubility in water: sparingly soluble
Formula Weight 151.25
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Adamantanamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Adamantanamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.