missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl

Product Code. 11304995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304995 1 g 1g
11314995 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304995 Supplier Thermo Scientific Alfa Aesar Supplier No. L18426.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59084-16-1 | C8H12ClNO2 | 189.639 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59084-16-1
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.639
MDL Number MFCD02094017
InChI Key OHCPVLJEAHBMEG-UHFFFAOYSA-N
Synonym 1-acetylisonipecotoyl chloride, n-acetylisonipecotoyl chloride, 1-acetyl-4-piperidinecarbonyl chloride, 1-acetyl isonipecotoyl chloride, acmc-20aoys, acetylisonipecotoyl chloride, 1acetylisonipecotoyl chloride, n-acetylisonipecotyl chloride, ksc274a6b, n-acetyl isonipecotoyl chloride
PubChem CID 100950
IUPAC Name 1-acetylpiperidine-4-carbonyl chloride
SMILES CC(=O)N1CCC(CC1)C(=O)Cl

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
UN Number UN3261
Beilstein 389571
Sensitivity Moisture sensitive
Formula Weight 189.64
Percent Purity 97%
Chemical Name or Material 1-Acetylpiperidine-4-carbonyl chloride, may contain up to ≈1M free HCl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.