missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carbonitrile, 97%, Thermo Scientific™

Product Code. p-4812580
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10603043 250 mg 250mg
10773843 1 g 1g
10472464 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10603043 Supplier Thermo Scientific Maybridge Supplier No. SEW04588CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25503-91-7
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.20
MDL Number MFCD00221055
InChI Key NFDGRMQIOHRQHF-UHFFFAOYSA-N
Synonym 1-acetyl-4-piperidinecarbonitrile, 1-acetyl-4-cyanopiperidine, 1-acetyl-isonipecotonitrile, 4-piperidinecarbonitrile,1-acetyl, 1-ethanoylpiperidine-4-carbonitrile
PubChem CID 2756286
IUPAC Name 1-acetylpiperidine-4-carbonitrile
SMILES CC(=O)N1CCC(CC1)C#N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 152.2
Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.