missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetylpiperazine, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 1-Acetylpiperazine |
| CAS | 13889-98-0 |
| Molecular Formula | C6 H12 N2 O |
| Quantity | 100 g |
| Synonym | 4-Acetylpiperazine, 1-(Piperazinyl)ethanone, 1-(Piperazine-1-yl)ethan-1-one, 1-(Piperazin-1-yl)ethanone, 1-(1-Piperazinyl)ethanone |
| InChI Formula | InChI=1S/C6H12N2O/c1-6(9)8-4-2-7-3-5-8/h7H,2-5H2,1H3 |
| SMILES | CC(=O)N1CCNCC1 |
| IUPAC Name | 1-piperazin-1-ylethanone |
| Molecular Weight (g/mol) | 128.172 |
| Formula Weight | 128.095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction