missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperazine, 98%

Product Code. 15453207
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Unit Size:
100g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453207 50 g 50g
11315299 5 g 5g
11325299 25 g 25g
11335299 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15453207 Supplier Thermo Scientific Alfa Aesar Supplier No. B20200.18
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13889-98-0 | C6H12N2O | 128.175 g/mol

1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

Solubility
Soluble in water 210 g/L (20°C).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13889-98-0
MDL Number MFCD00058676
Synonym 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PubChem CID 83795
IUPAC Name 1-piperazin-1-ylethanone

Specifications

Melting Point 30°C to 34°C
Boiling Point 127°C to 130°C (8 mmHg)
Flash Point >110°C
Molecular Formula C6H13N2O
Quantity 50 g
Beilstein 112220
Sensitivity Hygroscopic
Solubility Information Soluble aq. soln. 210 g/L (20°C).
InChI Key PKDPUENCROCRCH-UHFFFAOYSA-O
SMILES CC(=O)N1CC[NH2+]CC1
Molecular Weight (g/mol) 129.18
Formula Weight 128.18
Percent Purity 99%
Chemical Name or Material 1-Acetylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.