missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylisatin, 97%

Product Code. 11378536
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378536 1 g 1g
11388536 5 g 5g
11398536 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378536 Supplier Thermo Scientific Alfa Aesar Supplier No. L17597.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 574-17-4 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-17-4
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00158542
InChI Key LPGDEHBASRKTDG-UHFFFAOYSA-N
Synonym 1-acetylisatin, n-acetylisatin, 1-acetyl-1h-indole-2,3-dione, acetylisatin, 1-acetylindoline-2,3-dione, 1-acetyl-indole-2,3-dione, 1h-indole-2,3-dione, 1-acetyl, indole-2,3-dione, 1-acetyl, 1-acetyl-2,3-dihydro-1h-indole-2,3-dione, n-acetyl-isatin
PubChem CID 11321
ChEBI CHEBI:16050
IUPAC Name 1-acetylindole-2,3-dione
SMILES CC(=O)N1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Beilstein 113959
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 1-Acetylisatin
compliance-icons
Product Identifier
  • 1-Acetylisatin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Acetyl-1H-indole-2,3-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.