missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylindoline-5-sulfonyl chloride, 97%

Product Code. 15466947
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15466947

Brand: Thermo Scientific Alfa Aesar H34044.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52206-05-0 | C10H10ClNO3S | 259.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52206-05-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.70
MDL Number MFCD07368558
InChI Key QNFXLCHANYHGIF-UHFFFAOYSA-N
Synonym 1-acetyl-2,3-dihydro-1h-indole-5-sulfonyl chloride, 1-acetylindoline-5-sulfonyl chloride, 1-acetyl-5-indolinesulfonoyl chloride, 1h-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro, 1-acetyl-5-indolinesulfonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetyl-5-chlorosulfonyl indoline, 1-acetylindoline-5-sulfonylchloride, pubchem5449, acetylindolinesulfonoylchloride
PubChem CID 14024596
IUPAC Name 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride
SMILES CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O

Specifications

Melting Point 172°C to 174°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 259.71
Percent Purity 97%
Chemical Name or Material 1-Acetylindoline-5-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.