missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Product Code. 10022210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022210 100 g Glass bottle 100g
10683173 5 g Glass bottle 5g
10367490 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10022210 Supplier Thermo Scientific Acros Supplier No. 169501000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-imidazol-1-ylethanone
SMILES CC(=O)N1C=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Beige to White or Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf
Formula Weight 110.12
Percent Purity 98%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 1-Acetylimidazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.