missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl Isonipecotoyl Chloride, TRC

Product Code. 30319438
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319438 500 mg 500mg
30315209 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30319438 Supplier Toronto Research Chemicals Supplier No. TRCA179550500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Acetylisonipecotoyl Chloride
CAS 59084-16-1
Molecular Formula C8 H12 Cl N O2
Quantity 500 mg
Synonym 1-Acetyl-4-piperidinecarbonyl Chloride, 1-Acetyl-4-piperidylcarbonyl chloride, 1-Acetylisonipecotoyl chloride, 1-Acetylpiperidin-4-ylcarbonyl chloride, N-Acetylisonipecotoyl chloride
InChI Formula InChI=1S/C8H12ClNO2/c1-6(11)10-4-2-7(3-5-10)8(9)12/h7H,2-5H2,1H3
SMILES CC(=O)N1CCC(CC1)C(=O)Cl
IUPAC Name 1-acetylpiperidine-4-carbonyl chloride
Molecular Weight (g/mol) 189.64
Formula Weight 189.0557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.