missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-piperidone, 98%

Product Code. p-7021250
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11465613

Brand: Thermo Scientific Alfa Aesar A13512.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32161-06-1 | C7H11NO2 | 141.17 g/mol

1-Acetyl-4-piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs, and manufacture of fentanyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Acetyl-4-piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs, and manufacture of fentanyl.

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32161-06-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00006190
InChI Key NNFOVLFUGLWWCL-UHFFFAOYSA-N
Synonym 1-acetyl-4-piperidone, n-acetyl-4-piperidone, 4-piperidinone, 1-acetyl, 1-acetyl-4-piperidinone, 1-acetyl-piperidin-4-one, 1-acetyl-4-oxopiperidine, n-acetyl-4-piperidinone, n-acetyl4-piperidone, pubchem19615, 1-acetylpiperid-4-one
PubChem CID 122563
IUPAC Name 1-acetylpiperidin-4-one
SMILES CC(=O)N1CCC(=O)CC1

Specifications

Density 1.146
Boiling Point 218°C
Flash Point 112°C (234°F)
Refractive Index 1.503
Quantity 5 g
Beilstein 116031
Solubility Information Soluble in water. Insoluble in acetic acid.
Formula Weight 141.17
Percent Purity 99%
Chemical Name or Material 1-Acetyl-4-piperidone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.