missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%

Product Code. 15445847
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445847 5 g 5g
15438295 250 mg 250mg
15435847 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15445847 Supplier Thermo Scientific Alfa Aesar Supplier No. H33058.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84162-82-3 | C14H15F2NO2 | 267.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84162-82-3
Molecular Formula C14H15F2NO2
Molecular Weight (g/mol) 267.276
MDL Number MFCD00179440
InChI Key UGPWQEIGJWVJDR-UHFFFAOYSA-N
Synonym 4-2',4'-difluorobenzoyl-1-acetylpiperidine, 1-4-2,4-difluorobenzoyl piperidin-1-yl ethanone, 1-acetylpiperidin-4-yl 2,4-difluorophenyl methanone, 1-acetyl-4-2,4-difluorobenzoyl piperidine, 1-4-2,4-difluoro-benzoyl-piperidin-1-yl-ethanone, 1-acetyl-4-2',4'-difluorobenzoyl piperidine, 1-4-2,4-difluorobenzoyl-1-piperidinyl-1-ethanone, 1-4-2.4-difluoro-benzoyl-piperidin-1-yl-ethanone, maybridge3_006593
PubChem CID 2740707
IUPAC Name 1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)C(=O)C2=C(C=C(C=C2)F)F

Specifications

Melting Point 97°C to 98°C
Quantity 5 g
Formula Weight 267.28
Percent Purity 96%
Chemical Name or Material 1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.