missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-3-indolecarboxaldehyde, 98%, Thermo Scientific™

Product Code. 10215154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10215154 1 g 1g
10326494 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215154 Supplier Thermo Scientific Acros Supplier No. 399700010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22948-94-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00039691
InChI Key LCJLFGSKHBDOAY-UHFFFAOYSA-N
Synonym 1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1
PubChem CID 89915
IUPAC Name 1-acetylindole-3-carbaldehyde
SMILES CC(=O)N1C=C(C2=CC=CC=C21)C=O

Specifications

Melting Point 165°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 187.2
Percent Purity 98%
Chemical Name or Material 1-Acetyl-3-indolecarboxaldehyde
compliance-icons
Product Identifier
  • 1-Acetyl-3-indolecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.