missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-2-naphthol, 99%

Product Code. 11329609
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329609 5 g 5g
11408457 25 g 25g
11418457 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329609 Supplier Thermo Scientific Alfa Aesar Supplier No. A18537.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 574-19-6 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-19-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00012131
InChI Key VUIOUIWZVKVFCI-UHFFFAOYSA-N
Synonym 2'-hydroxy-1'-acetonaphthone, 1-acetyl-2-naphthol, 1-2-hydroxynaphthalen-1-yl ethanone, 1-2-hydroxy-1-naphthalenyl ethanone, 1-2-hydroxy-1-naphthyl ethan-1-one, 2-hydroxy-1-acetonaphthone, 1-2-hydroxy-1-naphthyl ethanone, ethanone, 1-2-hydroxy-1-naphthalenyl, 1-acetyl-2-hydroxynaphthalene, 1'-acetonaphthone, 2'-hydroxy
PubChem CID 68455
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)ethanone
SMILES CC(=O)C1=C(C=CC2=CC=CC=C21)O

Specifications

Melting Point 63°C to 67°C
Boiling Point 130°C to 132°C (2 mmHg)
Quantity 5 g
Beilstein 1869900
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 1-Acetyl-2-naphthol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.