missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylcyclohexene, 97%

Product Code. 10396110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396110 5 g Glass bottle 5g
10508100 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396110 Supplier Thermo Scientific Acros Supplier No. 102500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 932-66-1 | C8H12O | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-66-1
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00001547
InChI Key LTYLUDGDHUEBGX-UHFFFAOYSA-N
Synonym 1-acetyl-1-cyclohexene, 1-acetylcyclohexene, 1-cyclohex-1-en-1-yl ethanone, 1-cyclohexen-1-yl ethanone, ethanone, 1-1-cyclohexen-1-yl, ketone, 1-cyclohexen-1-yl methyl, unii-7539u6wqbl, methyl 1-cyclohexenyl ketone, 1-cyclohexen-1-yl methyl ketone, 1-1-cyclohexenyl-ethanone
PubChem CID 13612
IUPAC Name 1-(cyclohexen-1-yl)ethanone
SMILES CC(=O)C1=CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73°C
Color Yellow
Density 0.9600g/mL
Boiling Point 201°C to 202°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H9COCH3
Refractive Index 1.4900 to 1.492
Quantity 5 g
Beilstein 07, 58
Specific Gravity 0.96
Formula Weight 124.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Acetylcyclohexene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.