missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanedithiol, 95%

Product Code. 11337166
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11337166

Brand: Thermo Scientific Alfa Aesar L07172.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3489-28-9 | C9H20S2 | 192.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3489-28-9
Molecular Formula C9H20S2
Molecular Weight (g/mol) 192.38
MDL Number MFCD00004914
InChI Key GJRCLMJHPWCJEI-UHFFFAOYSA-N
Synonym 1,9-nonanedithiol, 1,9-dimercaptononane, nonamethylene dimercaptan, unii-5bpj1xqe3t, 5bpj1xqe3t, fema no. 3513, 1,9-nonane dithiol, 1,9-dinonyl mercaptan, acmc-20ap0g
PubChem CID 248488
IUPAC Name nonane-1,9-dithiol
SMILES SCCCCCCCCCS

Specifications

Density 0.953
Boiling Point 351°C to 352°C
Flash Point >100°C (212°F)
Refractive Index 1.4999
Quantity 5 g
Beilstein 1737568
Sensitivity Air Sensitive
Formula Weight 192.39
Percent Purity 97%
Chemical Name or Material 1,9-Nonanedithiol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.