missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,9-Nonanediol, TRC
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
Specifications
Specifications
| Chemical Name or Material | 1,9-Nonanediol |
| CAS | 3937-56-2 |
| Molecular Formula | C9 H20 O2 |
| Quantity | 50 g |
| Synonym | 1,9-Nonanediol (6CI','7CI','8CI','9CI','ACI), 1,9-Dihydroxynonane, Nonamethylene glycol, NSC 5416, α,ω-Nonanediol |
| InChI Formula | InChI=1S/C9H20O2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2 |
| SMILES | OCCCCCCCCCO |
| IUPAC Name | nonane-1,9-diol |
| Molecular Weight (g/mol) | 160.25 |
| Formula Weight | 160.1463 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction