missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanediol, TRC

Product Code. 30482152
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482152 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482152 Supplier Toronto Research Chemicals Supplier No. TRCN64937010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,9-Nonanediol
CAS 3937-56-2
Molecular Formula C9 H20 O2
Quantity 10 g
Synonym 1,9-Nonanediol (6CI','7CI','8CI','9CI','ACI), 1,9-Dihydroxynonane, Nonamethylene glycol, NSC 5416, α,ω-Nonanediol
InChI Formula InChI=1S/C9H20O2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2
SMILES OCCCCCCCCCO
IUPAC Name nonane-1,9-diol
Molecular Weight (g/mol) 160.25
Formula Weight 160.1463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.