missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanediol Diacrylate (>90%), TRC

Product Code. 30428702
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428702 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428702 Supplier Toronto Research Chemicals Supplier No. TRCN8873305G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,9-Nonanediol Diacrylate
CAS 107481-28-7
Molecular Formula C15 H24 O4
Quantity 5 g
Synonym 1,9-Bis(acryloyloxy)nonane, 1,9-Nonamethylene diacrylate, 1.9ND-A, A-NOD, A-NOD-N, EM 2203, FA 129A, FA 129AS, Fancryl FA 129AS, Fancryl FA 129SA
InChI Formula InChI=1S/C15H24O4/c1-3-14(16)18-12-10-8-6-5-7-9-11-13-19-15(17)4-2/h3-4H,1-2,5-13H2
SMILES C=CC(=O)OCCCCCCCCCOC(=O)C=C
IUPAC Name 9-prop-2-enoyloxynonyl prop-2-enoate
Molecular Weight (g/mol) 268.35
Formula Weight 268.1675
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.