missing translation for 'onlineSavingsMsg'
Learn More

1,9-Diphenyl-1,3,6,8-nonatetraen-5-one, 97+%, Thermo Scientific Chemicals

Product Code. 11458197
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458197 5 g 5g
11468197 25 g 25g
11478197 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458197 Supplier Thermo Scientific Alfa Aesar Supplier No. A17568.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-21-9
Molecular Formula C21H18O
Molecular Weight (g/mol) 286.374
MDL Number MFCD00004792
InChI Key RLJALOQFYHCJKG-XCBHXTLASA-N
Synonym acetone, dicinnamylidene, 1z,3e,6e,8e-1,9-diphenylnona-1,3,6,8-tetraen-5-one, 1,9-diphenylnona-1,3,6,8-tetraen-5-one, 1,3,6,8-nonatetraen-5-one, 1,9-diphenyl, 1,6,8-nonatetraen-5-one, 1,9-diphenyl, 1z,3e,6e,8e-1,9-diphenyl-5-nona-1,3,6,8-tetraenone
PubChem CID 6436875
IUPAC Name (1Z,3E,6E,8E)-1,9-diphenylnona-1,3,6,8-tetraen-5-one
SMILES C1=CC=C(C=C1)C=CC=CC(=O)C=CC=CC2=CC=CC=C2

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Formula Weight 286.37
Percent Purity 98%
Chemical Name or Material 1,9-Diphenyl-1,3,6,8-nonatetraen-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.