missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dichlorononane, 98%, Thermo Scientific™

Product Code. 15421839
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421839 5 g 5g
15431839 25 g 25g
15441839 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421839 Supplier Thermo Scientific Alfa Aesar Supplier No. H64561.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. Many organisms use 1,9-dichlorononane as a sole carbon source. Bacteria on incubation of sewage with 1,9-dichlorononane increased its subsequent capacity to dehalogenate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-99-8
Molecular Formula C9H18Cl2
Molecular Weight (g/mol) 197.143
MDL Number MFCD00001024
InChI Key JMGRNJZUQCEJDB-UHFFFAOYSA-N
Synonym nonane, 1,9-dichloro, 1,9-dichlornonane, nonane,9-dichloro, ksc449k9f, 1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
PubChem CID 13191
IUPAC Name 1,9-dichlorononane
SMILES C(CCCCCl)CCCCCl

Specifications

Color Colorless
Density 1.09 g/mL
Boiling Point 258°C to 262°C
Flash Point >112°C (234°F)
Linear Formula Cl(CH2)9Cl
Refractive Index 1.459
Quantity 5 g
Beilstein 1698729
Solubility Information Insoluble in water.
Formula Weight 197.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,9-Dichlorononane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.