missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%

Product Code. 11426877
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426877 50 g 50g
11416877 10 g 10g
11424210 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426877 Supplier Thermo Scientific Alfa Aesar Supplier No. A14324.18
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4549-33-1 | C9H18Br2 | 286.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-33-1
Molecular Formula C9H18Br2
Molecular Weight (g/mol) 286.051
MDL Number MFCD00000279
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

Density 1.41
Boiling Point 285°C to 288°C
Flash Point >110°C (230°F)
Linear Formula Br(CH2)9Br
Refractive Index 1.4965
Quantity 50 g
Beilstein 1737598
Formula Weight 286.06
Percent Purity 97%
Chemical Name or Material 1,9-Dibromononane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.