missing translation for 'onlineSavingsMsg'
Learn More

1,9-Diaminononane, 98%

Product Code. 11391487
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391487 25 g 25g
11381487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391487 Supplier Thermo Scientific Alfa Aesar Supplier No. L02043.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 646-24-2 | C9H22N2 | 158.289 g/mol

1,9-Diaminononane is used as an organic intermediate. It is also used to detect spermine and spermidine by matrix-assisted laser desorption/ionization combined with time-of-flight mass spectrometry ( MALDI-TOF MS) in foods and for the identification of apoptosis-related proteins by nano-flow ultra-performance liquid chromatography tandem mass-spectrometry (UPLC-MS/MS) after treatment with spermine and spermidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,9-Diaminononane is used as an organic intermediate. It is also used to detect spermine and spermidine by matrix-assisted laser desorption/ionization combined with time-of-flight mass spectrometry ( MALDI-TOF MS) in foods and for the identification of apoptosis-related proteins by nano-flow ultra-performance liquid chromatography tandem mass-spectrometry (UPLC-MS/MS) after treatment with spermine and spermidine.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-24-2
Molecular Formula C9H22N2
Molecular Weight (g/mol) 158.289
MDL Number MFCD00008251
InChI Key SXJVFQLYZSNZBT-UHFFFAOYSA-N
Synonym 1,9-diaminononane, 1,9-nonanediamine, nonamethylenediamine, 1,9-diaminonane, nonan-1,9-diamin, 1.9-diaminononane, acmc-1atpv, 1,9-nonamethylenediamine
PubChem CID 69534
IUPAC Name nonane-1,9-diamine
SMILES C(CCCCN)CCCCN

Specifications

Melting Point 36°C to 40°C
Boiling Point 258°C to 259°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3259
Beilstein 1737533
Formula Weight 158.29
Percent Purity 98%
Chemical Name or Material 1,9-Diaminononane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.