missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,9-Decadiene, 97%, Thermo Scientific Chemicals
204.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
It is widely used in the manufacturers and suppliers of pharmaceuticals and intermediates in the preparation of fine chemicals. 1,9-decadiene is also used as a starting material in ADMET polymerizations .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.75 g/mL |
Boiling Point | 169°C |
Flash Point | 42°C (107°F) |
Refractive Index | 1.4325 |
Quantity | 50 g |
UN Number | UN3295 |
Solubility Information | It is soluble in organic solvents. |
Formula Weight | 138.25 |
Percent Purity | 97% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'