missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanedithiol, 98%

Product Code. 11430578
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430578 25 g 25g
11410578 1 g 1g
11420578 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430578 Supplier Thermo Scientific Alfa Aesar Supplier No. B24693.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1191-62-4 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-62-4
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00003574
InChI Key PGTWZHXOSWQKCY-UHFFFAOYSA-N
Synonym 1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
PubChem CID 14493
IUPAC Name octane-1,8-dithiol
SMILES SCCCCCCCCS

Specifications

Density 0.96
Boiling Point 269°C to 270°C
Flash Point 113°C (235°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 25 g
Beilstein 1735431
Sensitivity Air Sensitive
Formula Weight 178.36
Percent Purity 98%
Chemical Name or Material 1,8-Octanedithiol
Show More Show Less
compliance-icons
Product Identifier
  • 1,8-Octanedithiol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,8-Octanedithiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.