missing translation for 'onlineSavingsMsg'
Learn More

1,8-Nonadiyne, 97%

Product Code. 11430238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430238 25 g 25g
11420238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430238 Supplier Thermo Scientific Alfa Aesar Supplier No. B23784.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2396-65-8 | C9H12 | 120.195 g/mol

It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from heavy metal powders and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-65-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008581
InChI Key DMOVPHYFYSASTC-UHFFFAOYSA-N
Synonym 1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 16969
IUPAC Name nona-1,8-diyne
SMILES C#CCCCCCC#C

Specifications

Melting Point -21°C
Density 0.799
Boiling Point 55°C to 56°C (13 mmHg)
Flash Point 41°C (105°F)
Odor Characteristic
Linear Formula HC≡C(CH2)5C≡CH
Refractive Index 1.449
Quantity 25 g
UN Number UN3295
Beilstein 1733443
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity 97%
Chemical Name or Material 1,8-Nonadiyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.