missing translation for 'onlineSavingsMsg'
Learn More

1,8-Nonadiene, 99%

Product Code. 15490388
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490388 1 g 1g
15410398 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490388 Supplier Thermo Scientific Alfa Aesar Supplier No. H53502.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4900-30-5 | C9H16 | 124.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4900-30-5
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00008669
InChI Key VJHGSLHHMIELQD-UHFFFAOYSA-N
Synonym 1,8-nonadiene, acmc-209kep
PubChem CID 78612
IUPAC Name nona-1,8-diene
SMILES C=CCCCCCC=C

Specifications

Density 0.74
Boiling Point 141°C to 142°C
Flash Point 26°C (79°F)
Linear Formula CH2=CH(CH2)5CH=CH2
Refractive Index 1.427
Quantity 1 g
UN Number UN3295
Formula Weight 124.23
Percent Purity 99%
Chemical Name or Material 1,8-Nonadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.