missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthoic Anhydride, TRC

Product Code. 30475698
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30475698 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30475698

Brand: Toronto Research Chemicals TRCN345300250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naphthalic anhydride
CAS 81-84-5
Molecular Formula C12 H6 O3
Quantity 250 g
Synonym Naphthalic anhydride (7CI,8CI), 1,8-Naphthalenedicarboxylic acid anhydride, 1,8-Naphthalenedicarboxylic anhydride, 1,8-Naphthalic acid anhydride, 1,8-Naphthalic anhydride, 1,8-Naphthanoic anhydride, 1,8-Naphthoic anhydride, NSC 5747, Protect, Protect (agrochemical)
InChI Formula InChI=1S/C12H6O3/c13-11-8-5-1-3-7-4-2-6-9(10(7)8)12(14)15-11/h1-6H
SMILES O=C1OC(=O)c2cccc3cccc1c23
Molecular Weight (g/mol) 198.17
Formula Weight 198.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.