missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalimide, 98%

Product Code. 11443118
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443118 10 g 10g
11453118 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443118 Supplier Thermo Scientific Alfa Aesar Supplier No. A18856.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-83-4 | C12H7NO2 | 197.193 g/mol

It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-83-4
Molecular Formula C12H7NO2
Molecular Weight (g/mol) 197.193
MDL Number MFCD00006920
InChI Key XJHABGPPCLHLLV-UHFFFAOYSA-N
Synonym 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn
PubChem CID 66491
IUPAC Name benzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2

Specifications

Melting Point 299°C to 303°C
Quantity 10 g
Beilstein 153150
Solubility Information Insoluble in water.
Formula Weight 197.2
Percent Purity 98%
Chemical Name or Material 1,8-Naphthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.