missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalic anhydride, 97%

Product Code. 10781545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10781545 2.5 kg 2.5kg
10466940 100 g 100g
10655752 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10781545 Supplier Thermo Scientific Acros Supplier No. 128140025
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 81-84-5 | C12H6O3 | 198.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 81-84-5
Molecular Formula C12H6O3
Molecular Weight (g/mol) 198.18
MDL Number MFCD00006925
InChI Key GRSMWKLPSNHDHA-UHFFFAOYSA-N
Synonym 1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
PubChem CID 6693
ChEBI CHEBI:82246
IUPAC Name 3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point >269.0°C
Color Beige to Brown
Flash Point 272°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 2.5 kg
Beilstein 17, 521
Fieser 01,713
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in acetic acid
Formula Weight 198.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 8-Naphthalic anhydride, 97%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 1,8-Naphthalic anhydride
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Faresætning
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.