missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Diiodooctane, 97+%, stab. with copper, Thermo Scientific Chemicals
72.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
50 g
Description
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).
Solubility
Miscible with methanol.
Notes
Light sensitive. Incompatible with water and strong bases.
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).
Solubility
Miscible with methanol.
Notes
Light sensitive. Incompatible with water and strong bases.
Specifications
Density | 1.84 |
Boiling Point | 167°C to 169°C (6 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.5635 |
Quantity | 25 g |
Beilstein | 1735437 |
Sensitivity | Light sensitive |
Synonym | octane, 1,8-diiodo, octamethylene diiodide, pubchem3851, octane,1,8-diiodo, acmc-1cljq, 1,8-bis iodanyl octane, 1,8-diiodooctane, stabilized with copper, stabilizedwithcopperchip, 1,8-diiodooctane, contains copper as stabilizer |
Solubility Information | Miscible with methanol. |
SMILES | C(CCCCI)CCCI |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'