missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diiodooctane, 97+%, stab. with copper

Product Code. 11309216
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309216 10 g 10g
11434210 25 g 25g
11404213 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309216 Supplier Thermo Scientific Alfa Aesar Supplier No. A10867.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24772-63-2 | C8H16I2 | 366.025 g/mol

1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

Solubility
Miscible with methanol.

Notes
Light sensitive. Incompatible with water and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24772-63-2
Molecular Formula C8H16I2
MDL Number MFCD00001106
InChI Key KZDTZHQLABJVLE-UHFFFAOYSA-N
PubChem CID 90605
IUPAC Name 1,8-diiodooctane

Specifications

Density 1.84
Boiling Point 167°C to 169°C (6 mmHg)
Flash Point >110°C
Refractive Index 1.5635
Quantity 10 g
Beilstein 1735437
Sensitivity Light sensitive
Synonym octane, 1,8-diiodo, octamethylene diiodide, pubchem3851, octane,1,8-diiodo, acmc-1 cljq, 1,8-bis iodanyl octane, 1,8-diiodooctane, stabilized with copper, stabilizedwithcopperchip, 1,8-diiodooctane, contains copper as stabilizer
Solubility Information Miscible with methanol.
SMILES ICCCCCCCCI
Molecular Weight (g/mol) 366.03
Formula Weight 366.03
Percent Purity ≥97%
Chemical Name or Material 1,8-Diiodooctane, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.