missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, TRC

Product Code. 30376437
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376437 1 g 1g
30387151 5 g -
30386229 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376437 Supplier Toronto Research Chemicals Supplier No. TRCD4257151G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
Molecular Weight (g/mol) 272.02
Synonym NSC 9820, Octamethylene Dibromide, alpha,ω-Dibromooctane
IUPAC Name 1,8-dibromooctane
SMILES BrCCCCCCCCBr

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H16Br2/c9-7-5-3-1-2-4-6-8-10/h1-8H2
Formula Weight 269.9619
Chemical Name or Material 1,8-Dibromooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.