missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, 98%

Product Code. 10070770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070770 50 mL Glass bottle 50mL
10326370 10 mL Glass bottle 10mL
11934491 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10070770 Supplier Thermo Scientific Acros Supplier No. 112880500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4549-32-0 | C8H16Br2 | 272.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
Molecular Weight (g/mol) 272.01
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane
SMILES C(CCCCBr)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 15.0°C to 16.0°C
Color Yellow
Density 1.4600g/mL
Boiling Point 270.0°C to 272.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br(CH2)8Br
Refractive Index 1.4981 to 1.5001
Quantity 50 mL
Beilstein 01,16
Specific Gravity 1.46
Solubility Information Solubility in water: immiscible
Formula Weight 272.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 8-Dibromooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.