missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

Catalog Number 11479673
Click to view available options
Quantity:
25 g
100 g
500 g
6674-22-2
C9H16N2
152.241
MFCD00006930
GQHTUMJGOHRCHB-UHFFFAOYSA-N
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
81184
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
C1CCC2=NCCCN2CC1
6674-22-2
C9H16N2
152.241
MFCD00006930
GQHTUMJGOHRCHB-UHFFFAOYSA-N
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
81184
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
C1CCC2=NCCCN2CC1

1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.
TRUSTED_SUSTAINABILITY
Melting Point -70°C
Density 1.02
pH 12.8
Boiling Point 261°C
Flash Point 116°C (240°F)
Odor Amine-like
Refractive Index 1.522
Quantity 25 g
UN Number UN3267
Beilstein 508906
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'