missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%

Product Code. 11489673
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489673 100 g 100g
11479673 25 g 25g
11499673 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489673 Supplier Thermo Scientific Alfa Aesar Supplier No. A12449.22
  • ONLINE EXCLUSIVE PRICE
    £63.40
    £57.65 / 100g
    Save £5.75
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6674-22-2 | C9H16N2 | 152.241 g/mol

1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6674-22-2
Molecular Formula C9H16N2
Molecular Weight (g/mol) 152.241
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

Melting Point -70°C
Density 1.02
pH 12.8
Boiling Point 261°C
Flash Point 116°C (240°F)
Odor Amine-like
Refractive Index 1.522
Quantity 100 g
UN Number UN3267
Beilstein 508906
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Diazabicyclo[5.4.0]undec-7-ene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,8-Diazabicyclo[5.4.0]undec-7-ene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.