missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaminooctane, 98%

Product Code. 10783455 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10783455 500 g Glass bottle 500g
10664952 25 g Glass bottle 25g
10122810 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10783455 Supplier Thermo Scientific Acros Supplier No. 112315000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 373-44-4 | C8H22Cl2N2 | 217.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 50.0°C to 54.0°C
Color White to Yellow
Density 165g/mL
Boiling Point 225.0°C to 226.0°C
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)8NH2
Quantity 500 g
Beilstein 04, 271
Solubility Information Solubility in water: 575g/L (20°C).
IUPAC Name octane-1,8-diamine
Formula Weight 144.26
Percent Purity 98%
Physical Form Solidified Crystalline Mass
Chemical Name or Material 1, 8-Diaminooctane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Diaminooctane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.