missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaminonaphthalene, 97%

Product Code. 10257543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10257543 2.5 kg Glass Bottle 2.5kg
10304082 5 g Glass Bottle 5g
10592072 100 g Glass Bottle 100g
10151292 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10257543 Supplier Thermo Scientific Acros Supplier No. 371180025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 479-27-6 | C10H10N2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-27-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004033
InChI Key YFOOEYJGMMJJLS-UHFFFAOYSA-N
Synonym 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
PubChem CID 68067
IUPAC Name naphthalene-1,8-diamine
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color Brown or Gray to Gray-Red
Boiling Point 205.0°C (12.0 mmHg)
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula C10H6(NH2)2
Quantity 2.5 kg
Beilstein 13, 205
Merck Index 15, 6457
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 158.2
Percent Purity 97%
Physical Form Crystalline Mass or Chunks
Chemical Name or Material 1, 8-Diaminonaphthalene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Diaminonaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.