missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Cineole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.
Solubility
Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.
Notes
Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.
Solubility
Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.
Notes
Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.
Specifications
Melting Point | 1°C |
Density | 0.92 |
Boiling Point | 176°C to 177°C |
Flash Point | 49°C (120°F) |
Assay Percent Range | 99% |
Refractive Index | 1.4575 |
Quantity | 100 mL |
UN Number | UN3271 |
Beilstein | 105109 |
Merck Index | 14,3895 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'