missing translation for 'onlineSavingsMsg'
Learn More

1,8-Cineole, 99%, Thermo Scientific Chemicals

Catalog Number 11444983
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
470-82-6
C10H18O
154.25
MFCD00167977
WEEGYLXZBRQIMU-UHFFFAOYSA-N
eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
2758
2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
CC12CCC(CC1)C(C)(C)O2
This item is not returnable. View return policy
470-82-6
C10H18O
154.25
MFCD00167977
WEEGYLXZBRQIMU-UHFFFAOYSA-N
eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
2758
2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
CC12CCC(CC1)C(C)(C)O2

Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

Solubility
Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.

Notes
Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.
TRUSTED_SUSTAINABILITY
Melting Point 1.0°C
Density 0.923
Boiling Point 176°C to 177°C
Flash Point 49°C (120°F)
Assay Percent Range 99%
Refractive Index 1.4575
Quantity 500 mL
UN Number UN3271
Beilstein 105109
Merck Index 14,3895
Solubility Information Soluble in water(3500mg/L (at 21°C). Miscible with ether,alcohol,chloroform,glacial acetic acid,oils. Soluble in ethanol,ethyl ether; slightly soluble in carbon tetrachloride.
Formula Weight 154.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'