missing translation for 'onlineSavingsMsg'
Learn More

1-(8-Chloro-11-(4-methylpiperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepin-5-yl)ethanone, TRC

Product Code. 30007414
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007414 50 mg 50mg
30003551 500 mg 500mg
30006101 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007414 Supplier Toronto Research Chemicals Supplier No. TRCC36755350MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025951-76-1
Molecular Formula C20H21ClN4O
Molecular Weight (g/mol) 368.86
Synonym 5-Acetyl-8-chloro-11-(4-methylpiperazin-1-yl)-5H-dibenzo-(b,e)(1,4)-diazepine, Clozapine Impurity, 1-[8-chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepin-5-yl]ethanone
IUPAC Name 1-[3-chloro-6-(4-methylpiperazin-1-yl)benzo[b][1,4]benzodiazepin-11-yl]ethanone
SMILES CN1CCN(CC1)C2=Nc3cc(Cl)ccc3N(C(=O)C)c4ccccc24

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H21ClN4O/c1-14(26)25-18-6-4-3-5-16(18)20(24-11-9-23(2)10-12-24)22-17-13-15(21)7-8-19(17)25/h3-8,13H,9-12H2,1-2H3
Formula Weight 368.1404
Chemical Name or Material 1-(8-Chloro-11-(4-methylpiperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepin-5-yl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.