missing translation for 'onlineSavingsMsg'
Learn More

1,7-Phenanthroline, 99%

Product Code. 10336494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336494 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10336494 Supplier Thermo Scientific Acros Supplier No. 426110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 230-46-6 | C12H8N2 | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 230-46-6
CAS Min % 98.5
CAS Max % 100.0
Melting Point 77°C to 78°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H8N2
MDL Number MFCD00004971
Quantity 1 g
Synonym pyridino 2,3-f quinoline, ccris 7840, 1,7 phenanthroline, acmc-20ap0u, opera_id_785, 1,7-phenanthroline
InChI Key OZKOMUDCMCEDTM-UHFFFAOYSA-N
SMILES C1=CN=C2C(C=CC3=NC=CC=C23)=C1
IUPAC Name 1,7-phenanthroline
Molecular Weight (g/mol) 180.21
PubChem CID 67473
ChEBI CHEBI:36418
Formula Weight 180.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 7-Phenanthroline
Show More Show Less
compliance-icons
Product Identifier
  • 1,7-Phenanthroline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.