missing translation for 'onlineSavingsMsg'
Learn More

1,7-Heptanediol, 95%

Product Code. p-7044433
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11382496

Brand: Thermo Scientific Alfa Aesar L06384.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-30-1 | C7H16O2 | 132.203 g/mol

1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water, slightly soluble in ether.

Notes
Incompatible with reducing agents, oxidizing agents, Acid chlorides, Acid anhydrides and chloroformates. Store away from acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-30-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.203
MDL Number MFCD00002987
InChI Key SXCBDZAEHILGLM-UHFFFAOYSA-N
Synonym 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor
PubChem CID 12381
IUPAC Name heptane-1,7-diol
SMILES C(CCCO)CCCO

Specifications

Melting Point 17°C to 18°C
Density 0.953
Boiling Point 258°C to 260°C
Flash Point 127°C (260°F)
Refractive Index 1.455
Quantity 5 g
Beilstein 1633482
Solubility Information Soluble in water,slightly soluble in ether.
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 1,7-Heptanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.