missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dioxaspiro[5.5]undecane, TRC

Product Code. 30382743
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
Product Code. Quantity unitSize
30382743 500 mg -
30372825 5 g -
30373505 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 30382743

Brand: Toronto Research Chemicals TRCD486325500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180-84-7
Formula Weight 156.115
InChI Formula InChI=1S/C9H16O2/c1-3-7-10-9(5-1)6-2-4-8-11-9/h1-8H2
IUPAC Name 1,7-dioxaspiro[5.5]undecane
Molecular Formula C9 H16 O2
Molecular Weight (g/mol) 156.22
SMILES C1CCC2(CCCCO2)OC1
Synonym 1,7-Dioxaspiro[5.5]undecane, Olean
Chemical Name or Material 1,7-Dioxaspiro[5.5]undecane

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.