missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dihydroxynaphthalene, 97%

Product Code. 11317649
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317649 25 g 25g
11307649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317649 Supplier Thermo Scientific Alfa Aesar Supplier No. H28261.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 575-38-2 | C10H8O2 | 160.172 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 575-38-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00035720
InChI Key ZUVBIBLYOCVYJU-UHFFFAOYSA-N
Synonym 1,7-dihydroxynaphthalene, 1,7-naphthalenediol, napthalene-1,7-diol, unii-7d42f605cs, ccris 7895, 1,7 dihydroxynaphthalene, 2,8-dihydroxynaphthalene, acmc-1aqz2, unii-m7uz7bfo3a component
PubChem CID 68462
IUPAC Name naphthalene-1,7-diol
SMILES C1=CC2=C(C=C(C=C2)O)C(=C1)O

Specifications

Melting Point 180°C to 184°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 1,7-Dihydroxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.