missing translation for 'onlineSavingsMsg'
Learn More

1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Product Code. 15497565
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15497565

Brand: Thermo Scientific Alfa Aesar H26293.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 162148-48-3 | C20H40N4O4 | 400.56 g/mol

Intermediate in the preparation of chelating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in the preparation of chelating agents.

Solubility
Soluble in acetone, acetonitrile, ethyl Acetate, chloroforme, cyclohexane. Insoluble in water.

Notes
Hygroscopic. Store in cool place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 162148-48-3
Molecular Formula C20H40N4O4
MDL Number MFCD09263303
Quantity 250 mg
Sensitivity Hygroscopic
Synonym di-tert-butyl 2,2'-1,4,7,10-tetraazacyclododecane-1,7-diyl diacetate, 1,7-bis tert-butoxycarbonylmethyl-1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane-1,7-diacetic acid 1,7-bis 1,1-dimethylethyl ester, tert-butyl 2-7-2-tert-butoxy-2-oxoethyl-1,4,7,10-tetraazacyclododecan-1-yl acetate, 1,4,7,10-tetraazacyclododecane-1,7-bis t-butyl acetate, 1,7-bis carbo-tert-butoxymethyl-1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane-1,7-bis acetic acid tert-butyl ester, 1,4,7,10-tetraazacyclododecane-1,7-diacetic acid, bis 1,1-dimethylethyl ester, tert-butyl 2-7-2-tert-butoxy-2-oxo-ethyl-1,4,7,10-tetrazacyclododec-1-yl acetate, tert-butyl 2-7-2-2-methylpropan-2-yl oxy-2-oxoethyl-1,4,7,10-tetrazacyclododec-1-yl acetate
Solubility Information Soluble in acetone,acetonitrile,ethyl Acetate,chloroforme,cyclohexane. Insoluble in water.
InChI Key OERWIIJOLSXGAQ-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCNCC1
IUPAC Name tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Molecular Weight (g/mol) 400.56
PubChem CID 393881
Formula Weight 400.56
Chemical Name or Material 1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.