missing translation for 'onlineSavingsMsg'
Learn More

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one oxime, 97%, Thermo Scientific™

Product Code. 10120582
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10120582 10 g 10g
10262532 1 g 1g
11320547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10120582 Supplier Thermo Scientific Maybridge Supplier No. BTB09928EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13559-66-5
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00210161
InChI Key OVFDEGGJFJECAT-DHZHZOJOSA-N
Synonym 2-bornanone oxime, camphor, d-oxime, 2-camphanone oxime, camphoroxime, bornan-2-one oxime, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one oxime, +-camphor oxime, camphor oxime, d-camphor oxime, camphor, oxime 6ci,7ci,8ci
PubChem CID 5908504
IUPAC Name (NE)-N-(4,7,7-trimethyl-3-bicyclo[2.2.1]heptanylidene)hydroxylamine
SMILES CC1(C2CCC1(C(=NO)C2)C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-one oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.